Anti-FXR1 Mouse Monoclonal Antibody [clone: 2G11]
USBI127036
- Antibody type:Primary
- Antigen name:Fragile X Mental Retardation 1 Protein
- Antigen symbol:FMR1
- Clonality:Monoclonal
- Clone:2G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:VKKNTFFKCTVDVPEDLREACANENAHKDFKKAVGACRIFYHPETTQLMILSASEATVKRVNILSDMHLRSIRTKLMLMSRNEEATKHLECTKQLAAAFH
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: FXR1 (fragile X mental retardation, autosomal homolog 1)
Clonality: monoclonal
Clone: 2G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human