Anti-ALB Rabbit Polyclonal Antibody
Catalog # 126705-1
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:albumin
- Antigen symbol:ALB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Chicken
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken