Anti-DYRK1A Mouse Monoclonal Antibody [clone: 7D10]
Catalog # USBI126095
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A
- Antigen symbol:DYRK1A
- Clonality:Monoclonal
- Clone:7D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:674 to 763
- Storage buffer:PBS, pH 7,2.
- Sequence:NQGNQAYQNRPVAANTLDFGQNGAMDVNLTVYSNPRQETGIAGHPTYQFSANTGPAHYMTEGHLTMRQGADREESPMTGVCVQQSPVASS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DYRK1A (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A)
Clonality: monoclonal
Clone: 7D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat