Anti-SLC44A2 Mouse Monoclonal Antibody [clone: 3D11]
USBI125445
- Antibody type:Primary
- Antigen name:solute carrier family 44 (choline transporter), member 2
- Antigen symbol:SLC44A2
- Clonality:Monoclonal
- Clone:3D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:YLNARSSRDFEYYKQFCVPGFKNNKGVAEVLRDGDCPAVLIPSKPLARRCFPAIHAYKGVLMVGNETTYEDGHGSRKNITDLVEGAKKANGVLEARQLAMRIFEDYTV
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: SLC44A2 (solute carrier family 44 (choline transporter), member 2)
Clonality: monoclonal
Clone: 3D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human