Anti-CCL13 Mouse Polyclonal Antibody
Catalog # USBI124440
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 13
- Antigen symbol:CCL13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 98
- Storage buffer:PBS, pH 7,2.
- Sequence:MKVSAVLLCLLLMTAAFNPQGLAQPDALNVPSTCCFTFSSKKISLQRLKSYVITTSRCPQKAVIFRTKLGKEICADPKEKWVQNYMKHLGRKAHTLKT
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CCL13 (chemokine (C-C motif) ligand 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human