Anti-CA7 Mouse Monoclonal Antibody [clone: 3B7]
Catalog # USBI124191
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:carbonic anhydrase VII
- Antigen symbol:CA7
- Clonality:Monoclonal
- Clone:3B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:34 to 125
- Storage buffer:PBS, pH 7,2.
- Sequence:NIISSQAVYSPSLQPLELSYEACMSLSITNNGHSVQVDFNDSDDRTVVTGGPLEGPYRLKQFHFHWGKKHDVGSEHTVDGKSFPSELHLVH*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: CA7 (carbonic anhydrase VII)
Clonality: monoclonal
Clone: 3B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human