Anti-CA5B Mouse Monoclonal Antibody [clone: 1E12]
USBI124188
- Antibody type:Primary
- Antigen name:carbonic anhydrase VB, mitochondrial
- Antigen symbol:CA5B
- Clonality:Monoclonal
- Clone:1E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MVVMNSLRVILQASPGKLLWRKFQIPRFMPARPCSLYTCTYKTRNRALHPLWESVDLVPGGDRQSPINIRWRDSVYDPGLKPLTISYDPATCLHVWNNGYSFLVEFEDSTDKSVIKGGPLEHNYRLKQFHFHWGAIDAWGSEHTVDSKCFPAELHLVHWNAVRFENFEDAALEENGLAVIGVFLKLGKHHKELQKLVDTLPSIKHKDALVEFGSFDPSCLMPTCPDYWTYSGSLTTPPLSESVTWIIKKQPVEVD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CA5B (carbonic anhydrase VB, mitochondrial)
Clonality: monoclonal
Clone: 1000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human