Anti-CA10 Mouse Monoclonal Antibody [clone: 1A10]
Catalog # USBI124178
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase X
- Antigen symbol:CA10
- Clonality:Monoclonal
- Clone:1A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:229 to 329
- Storage buffer:PBS, pH 7,2.
- Sequence:TSSFITYDGSMTIPPCYETASWIIMNKPVYITRMQMHSLRLLSQNQPSQIFLSMSDNFRPVQPLNNRCIRTNINFSLQGKDCPNNRAQKLQYRVNEWLLK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CA10 (carbonic anhydrase X)
Clonality: monoclonal
Clone: 1A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human