Anti-C1QTNF9 Mouse Polyclonal Antibody
USBI124111
- Antibody type:Primary
- Antigen name:C1q and tumor necrosis factor related Protein 9
- Antigen symbol:C1QTNF9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: C1QTNF9 (C1q and tumor necrosis factor related protein 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human