Anti-DEFB104A Mouse Monoclonal Antibody [clone: 4C4]
Catalog # USBI123906
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:defensin, beta 104A
- Antigen symbol:DEFB104A
- Clonality:Monoclonal
- Clone:4C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:24 to 64
- Storage buffer:PBS, pH 7,2.
- Sequence:SandwichELISA:Thedetectionlimitis~3ng/mlasacaptureantibody
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DEFB104A (defensin, beta 104A)
Clonality: monoclonal
Clone: 4C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human