Anti-SAMD10 Rabbit Polyclonal Antibody
Catalog # MEDMMM-0226
Supplier: MédiMabs
Specifications
- Antibody type:Primary
- Antigen name:Sterile alpha motif domain containing 10
- Antigen symbol:SAMD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SAMD10 (sterile alpha motif domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse