Anti-MAPT Mouse Monoclonal Antibody
Catalog # MEDMMM-0194-P
Supplier: MédiMabs
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein tau
- Antigen symbol:MAPT
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat