Anti-Cdc5 (Msd2-77) Mouse Monoclonal Antibody [clone: 4F10-11H12]
MEDMMM-0192-3-100
: MédiMabs
- Antibody type:Primary
- Antigen name:Msd2-77
- Antigen symbol:CDC5
- Clonality:Monoclonal
- Clone:4F10-11H12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Cdc5 (Msd2-77)
Clonality: monoclonal
Clone: 4F10-11H12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: