Order Entry
Italy
ContactUsLinkComponent
Anti-Cdc5 (Msd2-77) Mouse Monoclonal Antibody [clone: 11H12]
Anti-Cdc5 (Msd2-77) Mouse Monoclonal Antibody [clone: 11H12]
Catalog # MEDMMM-0192-2-100
Supplier:  MédiMabs
Anti-Cdc5 (Msd2-77) Mouse Monoclonal Antibody [clone: 11H12]
Catalog # MEDMMM-0192-2-100
Supplier:  MédiMabs

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Msd2-77
  • Antigen symbol:
    CDC5
  • Clonality:
    Monoclonal
  • Clone:
    11H12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Cdc5 (Msd2-77)
Clonality: monoclonal
Clone: 11H12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: