Anti-CHGA Rabbit Polyclonal Antibody
MEDMMM-0179
: MédiMabs
- Antibody type:Primary
- Antigen name:Chromogranin A
- Antigen symbol:CHGA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: CHGA (chromogranin A (parathyroid secretory protein 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: