Anti-GLIPR1L1 Rabbit Polyclonal Antibody
Catalog # MEDMMM-0162
Supplier: MédiMabs
Specifications
- Antibody type:Primary
- Antigen name:GLI pathogenesis-related 1 like 1
- Antigen symbol:GLIPR1L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Bovine
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GLIPR1L1 (GLI pathogenesis-related 1 like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine