Anti-SMAP2 Rabbit Polyclonal Antibody
Catalog # MEDMMM-0139
Supplier: MédiMabs
Specifications
- Antibody type:Primary
- Antigen name:Small ArfGAP 2
- Antigen symbol:SMAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SMAP2 (small ArfGAP2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human