Order Entry
Italy
ContactUsLinkComponent
 
Anti-RAB27A Rabbit Polyclonal Antibody
  MEDMMM-0132
 :  MédiMabs
Anti-RAB27A Rabbit Polyclonal Antibody
  MEDMMM-0132
 :  MédiMabs

 

  • Antibody type:
    Primary
  • Antigen name:
    RAB27A, member RAS oncogene family
  • Antigen symbol:
    RAB27A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: RAB27A (RAB27A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human