Anti-UNC13D Rabbit Polyclonal Antibody
Catalog # MEDMMM-0130
Supplier: MédiMabs
Specifications
- Antibody type:Primary
- Antigen name:Unc-13 homolog D (C. elegans)
- Antigen symbol:UNC13D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: UNC13D (unc-13 homolog D (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human