Order Entry
Italy
ContactUsLinkComponent
Anti-CLC Rabbit polyclonal antibody unconjugated
Anti-CLC Rabbit polyclonal antibody unconjugated
Catalog # MEDMMM-0033
Supplier:  MédiMabs
Anti-CLC Rabbit polyclonal antibody unconjugated
Catalog # MEDMMM-0033
Supplier:  MédiMabs

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Charcot-Leyden crystal galectin
  • Antigen symbol:
    CLC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: CLC (Charcot-Leyden crystal galectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat