Order Entry
Italy
ContactUsLinkComponent
Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 17-142F]
Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 17-142F]
Catalog # USBI044902
Supplier:  US Biological
Anti-IL17A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 17-142F]
Catalog # USBI044902
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 17A
  • Antigen symbol:
    IL17A
  • Clonality:
    Monoclonal
  • Clone:
    17-142F
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Form:
    liquid
  • Storage buffer:
    0,2um-filtered PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: Monoclonal
Clone: 17-142F
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: