Order Entry
Italy
ContactUsLinkComponent
Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: Kairos-37]
Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: Kairos-37]
Catalog # USBI044509
Supplier:  US Biological
Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: Kairos-37]
Catalog # USBI044509
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Angiopoietin-like 3
  • Antigen symbol:
    Aminomethyltransferase
  • Clonality:
    Monoclonal
  • Clone:
    Kairos-37
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    ANGPTL3
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 1:1000-1:5000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: ANGPTL3 (angiopoietin-like 3)
Clonality: Monoclonal
Clone: Kairos-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: