Order Entry
Italy
ContactUsLinkComponent
 
Anti-UBE4A Rabbit Polyclonal Antibody
  USBI043540
undefined
Anti-UBE4A Rabbit Polyclonal Antibody
  USBI043540

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitination Factor E4A
  • Antigen symbol:
    UBE4A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    457 to 486
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: UBE4A (ubiquitination factor E4A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human