Anti-TSC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI043347-FITC
- Antibody type:Primary
- Antigen name:tuberous sclerosis 2
- Antigen symbol:TSC2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser1348
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot, Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!
Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse