Anti-TRIM69 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI043261-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 69
- Antigen symbol:TRIM69
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:108 to 137
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: TRIM69 (tripartite motif containing 69)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse