Order Entry
Italy
ContactUsLinkComponent
Anti-SERPINI1 Rabbit Polyclonal Antibody (Biotin)
Anti-SERPINI1 Rabbit Polyclonal Antibody (Biotin)
  USBI041591-BIOTIN
undefined
Anti-SERPINI1 Rabbit Polyclonal Antibody (Biotin)
  USBI041591-BIOTIN

 

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade I, member 1
  • Antigen symbol:
    SERPINI1
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    26 to 53
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: SERPINI1 (serpin peptidase inhibitor, clade I (neuroserpin), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human