Anti-LGR5, loop2 Rabbit Polyclonal Antibody
USBI037760
- Antibody type:Primary
- Antigen name:LGR5, loop2
- Antigen symbol:LGR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:689 to 719
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Immunofluorescence: 1:100; Flow Cytometry: 1:10-1:50; Western Blot: 1:1000
Type: Primary
Antigen: LGR5, loop2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse