Anti-LC3B Rabbit Polyclonal Antibody
200 µl
USBI037701
Recommended dilution: ELISA: 1:1000; Dot Blot 1:500
Type: Primary
Antigen: LC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Size:200 µl
- Clonality:Polyclonal
- Modification:phospho-Thr93
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Cat. No.:USBI037701
- Reactivity:Human
- Conjugation:Unconjugated
- Tested applications:Dot Blot
- Antigen symbol:LC3B
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- ELISA:Yes



