Anti-Interleukin-17 Receptor A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI036981-AP
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:Interleukin-17 Receptor A
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:720 to 747
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifiche
Informazioni su questo articolo
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: Interleukin-17 Receptor A
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse