Anti-FIG4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI035655-FITC
- Antibody type:Primary
- Antigen name:FIG4 homolog, SAC1 lipid phosphatase domain containing (S. cerevisiae)
- Antigen symbol:FIG4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Amino acid number:845 to 874
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: FIG4 (FIG4 homolog, SAC1 lipid phosphatase domain containing (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse