Anti-ESR1 isoform1 Rabbit Polyclonal Antibody
Catalog # USBI035233
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ESR1 isoform1
- Antigen symbol:ESR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:452 to 480
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:10-1:50
Type: Primary
Antigen: ESR1 isoform1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human