Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # USBI034210
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CREB binding protein
- Antigen symbol:CREBBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Amino acid number:169 to 199
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000
Type: Primary
Antigen: CREBBP (CREB binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human