Anti-APOH Rabbit Polyclonal Antibody
USBI031990
- Antibody type:Primary
- Antigen name:Apolipoprotein H
- Antigen symbol:APOH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50
Type: Primary
Antigen: APOH (apolipoprotein H (beta-2-glycoprotein I))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human