Order Entry
Italy
ContactUsLinkComponent
Anti-MLST8 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 211CT7.1.1]
Anti-MLST8 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 211CT7.1.1]
Catalog # USBI030985-AP
Supplier:  US Biological
Anti-MLST8 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 211CT7.1.1]
Catalog # USBI030985-AP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MTOR associated Protein, LST8 homolog (S. cerevisiae)
  • Antigen symbol:
    MLST8
  • Clonality:
    Monoclonal
  • Clone:
    211CT7.1.1
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:1560

Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse