Order Entry
Italy
ContactUsLinkComponent
Anti-Microglobulin, pI 5.3 Mouse Monoclonal Antibody [clone: 467CT12.3.1]
Anti-Microglobulin, pI 5.3 Mouse Monoclonal Antibody [clone: 467CT12.3.1]
Catalog # USBI030798
Supplier:  US Biological
undefined
Anti-Microglobulin, pI 5.3 Mouse Monoclonal Antibody [clone: 467CT12.3.1]
Catalog # USBI030798
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Microglobulin, pI 5.3
  • Clonality:
    Monoclonal
  • Clone:
    467CT12.3.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-16000

Type: Primary
Antigen: Microglobulin, pI 5.3
Clonality: Monoclonal
Clone: 467CT12.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human