Order Entry
Italy
ContactUsLinkComponent
Anti-ATG7 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 29CT10.3.12]
Anti-ATG7 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 29CT10.3.12]
Catalog # USBI030780-AP
Supplier:  US Biological
Anti-ATG7 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 29CT10.3.12]
Catalog # USBI030780-AP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Autophagy related 7
  • Antigen symbol:
    APG7L
  • Clonality:
    Monoclonal
  • Clone:
    29CT10.3.12
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    APG7-like|ATG7
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-1000

Type: Primary
Antigen: ATG7 (autophagy related 7)
Clonality: Monoclonal
Clone: 29CT10.3.12
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human