Anti-A2M Mouse Monoclonal Antibody [clone: 590CT15.5.5]
Catalog # USBI030733
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Alpha-2-macroglobulin
- Antigen symbol:A2M
- Clonality:Monoclonal
- Clone:590CT15.5.5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-terminal
- Format:ascites
- Form:liquid
- Antigen synonyms:CPAMD5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-200
Type: Primary
Antigen: A2M (alpha-2-macroglobulin)
Clonality: Monoclonal
Clone: 590CT15.5.5
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Mouse