Anti-NFKBIB Mouse Monoclonal Antibody [clone: 13B569]
USBI030491
- Antibody type:Primary
- Antigen name:Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta
- Antigen symbol:NFKBIB
- Clonality:Monoclonal
- Clone:13B569
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: NFKBIB (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta)
Clonality: Monoclonal
Clone: 13B569
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human