Anti-RELA Mouse Monoclonal Antibody [clone: 13B566]
Catalog # USBI030488
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor p65
- Antigen symbol:RELA
- Clonality:Monoclonal
- Clone:13B566
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: Monoclonal
Clone: 13B566
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse