Anti-IL18 Rabbit Polyclonal Antibody
Catalog # USBI029817
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 18
- Antigen symbol:IL18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 1% BSA, 0,02% thimerosal, 50%glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-4?g/ml; Neutralization: 2-5 ?g/ml; Immunoprecipitation: 2-5 ?g/ml
Type: Primary
Antigen: IL18 (interleukin 18 (interferon-gamma-inducing factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse