Order Entry
Italy
ContactUsLinkComponent
Anti-SMARCE1 Rabbit Polyclonal Antibody
Anti-SMARCE1 Rabbit Polyclonal Antibody
Catalog # USBI029424
Supplier:  US Biological
Anti-SMARCE1 Rabbit Polyclonal Antibody
Catalog # USBI029424
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
  • Antigen symbol:
    SMARCE1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5 ?g/ml; Immunochemistry: 1:300

Type: Primary
Antigen: SMARCE1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat