Order Entry
Italy
ContactUsLinkComponent
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 53-6.7]
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 53-6.7]
Catalog # ROCK200-502-N77
undefined
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 53-6.7]
Catalog # ROCK200-502-N77

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    12525
  • Antigen synonyms:
    BB154331|Ly-2|Lyt-2|Ly-35|Ly-B
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD8a Antibody (Monoclonal) was produced by repeated immunizations with CD8a antigen.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Anti-CD8a is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse