Anti-HTT Rabbit Polyclonal Antibody
Catalog # ROCK600-401-433
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:huntingtin
- Antigen symbol:HTT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:3064
- Antigen synonyms:HD|IT15
- Amino acid number:416 to 424
- Modification:phospho-Ser421
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Huntingtin pS421 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 416-424 of Human huntingtin Protein.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-Huntingtin pS421 antibody has been tested for use in ELISA, immunochemistry and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect bands at approximately 350 kDa and 200 kDa in size.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:50 - 1:100;
Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human