Order Entry
Italy
ContactUsLinkComponent
Anti-HMGN1 Mouse Polyclonal Antibody (Unconjugated) [clone: IPO-38]
Anti-HMGN1 Mouse Polyclonal Antibody (Unconjugated) [clone: IPO-38]
Catalog # ROCK200-301-278
undefined
Anti-HMGN1 Mouse Polyclonal Antibody (Unconjugated) [clone: IPO-38]
Catalog # ROCK200-301-278

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    high mobility group nucleosomal binding domain 1
  • Antigen symbol:
    HMGN1
  • Clonality:
    Monoclonal
  • Clone:
    IPO-38
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    15312
  • Antigen synonyms:
    Hmg14|HMG-14
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    A BALB/c mouse was immunized with spleen cells of a human patient with hairy cell leukemia.
  • Purification:
    Protein A purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-IPO-38 is suitable for ELISA,  immunoprecipitation, western blotting and immunochemistry (frozen and formalin/paraffin). The antibody is reported to recognize a nuclear antigen that is present in the cytoplasm and nuclei of proliferating cells.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone: IPO-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse