Anti-MBP Mouse Monoclonal Antibody
Catalog # QEDB24201
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:myelin basic Protein
- Antigen symbol:MBP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: