Order Entry
Italy
ContactUsLinkComponent
Anti-20S Proteasome Core Subunits Rabbit Polyclonal Antibody
Anti-20S Proteasome Core Subunits Rabbit Polyclonal Antibody
  ST1053-100
Anti-20S Proteasome Core Subunits Rabbit Polyclonal Antibody
  ST1053-100

 

  • Antibody type:
    Primary
  • Antigen name:
    20S Proteasome Core Subunits
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: 20S Proteasome Core Subunits
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: