Order Entry
Italy
ContactUsLinkComponent
Anti-Acetylated Lysine Rabbit Polyclonal Antibody
Anti-Acetylated Lysine Rabbit Polyclonal Antibody
  ST1027-50
Anti-Acetylated Lysine Rabbit Polyclonal Antibody
  ST1027-50

 

  • Antibody type:
    Primary
  • Antigen name:
    Acetyl-Lysine
  • Antigen symbol:
    AcK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: Acetylated Lysine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan