Anti-FGF, Basic (Ab-2) (40-63) Rabbit Polyclonal Antibody
PC16-100
- Antibody type:Primary
- Antigen symbol:FGF basic
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Amino acid number:40 to 63
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: FGF, Basic (Ab-2) (40-63)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: