Order Entry
Italy
ContactUsLinkComponent
Anti-c-H-Ras (Ab-1) (F235-1.7.1) Mouse Monoclonal Antibody [clone: F235-1.7.1]
Anti-c-H-Ras (Ab-1) (F235-1.7.1) Mouse Monoclonal Antibody [clone: F235-1.7.1]
Catalog # OP23-100
Anti-c-H-Ras (Ab-1) (F235-1.7.1) Mouse Monoclonal Antibody [clone: F235-1.7.1]
Catalog # OP23-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    c-H-Ras
  • Clonality:
    Monoclonal
  • Clone:
    F235-1.7.1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: c-H-Ras (Ab-1) (F235-1.7.1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: