Order Entry
Italy
ContactUsLinkComponent
Anti-c-Abl (Ab-3) (24-21) Mouse Monoclonal Antibody [clone: 24-21]
Anti-c-Abl (Ab-3) (24-21) Mouse Monoclonal Antibody [clone: 24-21]
Catalog # OP20-100
Anti-c-Abl (Ab-3) (24-21) Mouse Monoclonal Antibody [clone: 24-21]
Catalog # OP20-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    C-ABL
  • Clonality:
    Monoclonal
  • Clone:
    24-21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: c-Abl (Ab-3) (24-21)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: